Learn More
CTSK Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CTSK.
Supplier: Abnova Corporation PAB19139
Description
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predominantly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing. [provided by RefSeq
Specifications
CTSK | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant CTSK. | |
In 20mM Na-phosphate, 150mM NaCl, pH 7.5 | |
CTS02/CTSO/CTSO1/CTSO2/MGC23107/PKND/PYCD | |
Rabbit | |
IgG purification | |
RUO | |
1513 | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western blot (1:1000-1:3000) Immunoprecipitation (1:30-1:100) The optimal working dilution should be determined by the end user. | |
CTSK | |
CTSK | |
Recombinant protein corresponding to human CTSK. | |
100 μg | |
Primary | |
No cross reactivity. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only