Learn More
CXCR1/IL-8RA Rabbit anti-Mouse, R-PE, Clone: 1122A, R&D Systems
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB8628P025
Catalog No. FAB8628P025
Add to cart
This item is not returnable. View return policy
Description
Description
CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCR1/IL-8RA
1122A
Flow Cytometry 10 uL/10^6 cells
NP_839972
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 RA, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Rabbit
IgG
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse CXCR1 transfectants but not CXCR2 transfectants in flow cytometry.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant
CXCR1
A peptide corresponding to the N-terminal amino acid sequence of mouse CXCR1 Accession # NP_839972
Protein A or G purified from cell culture supernatant
RUO
Primary
3577
Mouse