Learn More
CXCR4 Rabbit anti-Human, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of CXCR4.
Manufacturer: Abnova Corporation PAB26089
Description
This gene encodes a CXC chemokine receptor specific for stromal cell-derived factor-1. The protein has 7 transmembrane regions and is located on the cell surface. It acts with the CD4 protein to support HIV entry into cells and is also highly expressed in breast cancer cells. Mutations in this gene have been associated with WHIM (warts, hypogammaglobulinemia, infections, and myelokathexis) syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
CXCR4 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7 ug/mL) The optimal working dilution should be determined by the end user. | |
CXCR4 | |
Liquid | |
CXCR4 | |
A synthetic peptide corresponding to 16 amino acids at N-terminus of human CXCR4. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of CXCR4. | |
In PBS (0.09% sodium azide) | |
P61073 | |
CD184|D2S201E|FB22|HM89|HSY3RR|LAP3|LCR1|LESTR|NPY3R|NPYR|NPYRL|NPYY3R|WHIM | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
7852 | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only