Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1357420.1ML

Catalog No. NBP13574201


Add to cart

Description

Description

Cyr61/CCN1 Polyclonal antibody specifically detects Cyr61/CCN1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cyr61/CCN1
Unconjugated
10mM PBS (pH 7.4) with Sodium Azide
CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
This product is specific for Cyr61
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50
Affinity Purified
CYR61
A synthetic peptide corresponding to internal region of human CYR61
Immunogen affinity purified
RUO
Primary
3491
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit