Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Cytokeratin 5/6, Clone: T16-K, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236738

Catalog No. NBP236738


Add to cart

Description

Description

Cytokeratin 5/6 Monoclonal antibody specifically detects Cytokeratin 5/6 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin 5/6
T16-K
Immunohistochemistry-Paraffin 1:100-1:200
Supernatant
KRT5
Peptide derived from C-terminal sequence of human cytokeratin 6. Antibody recognizes the epitope (identical with cytokeratin 5) between Thr548 - His564.
Tissue culture supernatant
RUO
Primary
3852
Human
Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Monoclonal
Human P13647 (CK5), P02538 (CK6)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only