Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
daf-3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of daf-3.
Supplier: Abnova Corporation PAB12557
Specifications
daf-3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of daf-3. | |
In antiserum (0.05% sodium azide) | |
O17532 | |
Rabbit | |
100 μL | |
Primary | |
Nematode | |
Liquid |
ChIP Assay, Western Blot | |
Unconjugated | |
Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
daf-3 | |
daf-3 | |
A synthetic peptide corresponding to amino acids 700-796 of Caenorhabditis elegans daf-3. | |
RUO | |
180431 | |
Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction