Learn More
dihydrotestosterone Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against dihydrotestosterone.
Manufacturer:  Abnova Corporation PAB5165
Catalog No. 89-138-580
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
dihydrotestosterone
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
5a-ANDROSTAN-17b-OL-3-ONE3-O-CARBOXYMETHYLOXIME.
RUO
Primary
Not Provided
ELISA, Immunoprecipitation
Rabbit polyclonal antibody raised against dihydrotestosterone.
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Rabbit
150 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal