Learn More
Dio3 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Dio3.
Manufacturer:  Abnova Corporation PAB12111
Catalog No. 89-115-609
Add to cart
Description
Description
Specifications
Specifications
Dio3
Unconjugated
Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Dio3
Liquid
Rabbit
100 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Murine, Rat
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Dio3.
In PBS (30% glycerol, 0.09% sodium azide)
P49897
Dio3
A synthetic peptide corresponding to amino acids 1-100 of rat Dio3.
RUO
Primary
29475