Learn More
DISC1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DISC1.
Supplier: Abnova Corporation PAB5320
Description
This gene encodes a protein with multiple coiled coil motifs which is located in the nucleus, cytoplasm and mitochondria. The protein is involved in neurite outgrowth and cortical development through its interaction with other proteins. This gene is disrupted in a t(1;11)(q42.1;q14.3) translocation which segregates with schizophrenia and related psychiatric disorders in a large Scottish family. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
DISC1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of DISC1. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
C1orf136/FLJ13381/FLJ21640/FLJ25311/FLJ41105/KIAA0457/SCZD9 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
DISC1 | |
DISC1 | |
A synthetic peptide corresponding to N-terminus of human DISC1. | |
RUO | |
27185 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only