Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNM1L Rabbit anti-Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of DNM1L.

Manufacturer:  Abnova Corporation PAB12446

Catalog No. 89-115-820


Add to cart

Description

Description

The protein encoded by this gene is a member of the dynamin superfamily of GTPases. Members of the dynamin-related subfamily, including the S. cerevisiae proteins Dnm1 and Vps1, contain the N-terminal tripartite GTPase domain but do not have the pleckstrin homology or proline-rich domains. This protein establishes mitochondrial morphology through a role in distributing mitochondrial tubules throughout the cytoplasm. The gene has 3 alternatively spliced transcripts encoding different isoforms. These transcripts are alternatively polyadenylated. [provided by RefSeq

Specifications

Specifications

DNM1L
Unconjugated
Western Blot (1:500) The optimal working dilution should be determined by the end user.
DNM1L
Liquid
DNM1L
A synthetic peptide corresponding to amino acids 1-100 of human DNM1L.
RUO
Primary
10059
Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of DNM1L.
In Tris-glycine, 150mM NaCl (0.05% sodium azide)
O00429
DLP1|DRP1|DVLP|DYMPLE|DYNIV-11|FLJ41912|HDYNIV|VPS1
Rabbit
100 uL
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Primate, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only