Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of DNMT1.

Manufacturer:  Abnova Corporation PAB1833

Catalog No. 89-119-895


Add to cart

Description

Description

DNA (cytosine-5-)-methyltransferase 1 has a role in the establishment and regulation of tissue-specific patterns of methylated cytosine residues. Aberrant methylation patterns are associated with certain human tumors and developmental abnormalities. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Specifications

DNMT1
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
DNMT1
Liquid
DNMT1
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DNMT1.
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of DNMT1.
In PBS (0.09% sodium azide)
NP_001370;P26358
AIM|CXXC9|DNMT|FLJ16293|MCMT|MGC104992
Rabbit
Protein G purification
RUO
Primary
1786
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only