Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DOK6.
Supplier: Abnova Corporation PAB26918
Description
DOK6 is a member of the DOK (see DOK1; MIM 602919) family of intracellular adaptors that play a role in the RET (MIM 164761) signaling cascade (Crowder et al., 2004 [PubMed 15286081]).[supplied by OMIM
Specifications
DOK6 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
DOK6 | |
DOK5L/HsT3226/MGC20785 | |
Rabbit | |
Affinity purification | |
RUO | |
220164 | |
Human, Mouse | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of DOK6. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
Q6PKX4 | |
DOK6 | |
A synthetic peptide corresponding to human DOK6. | |
100 μL | |
Primary | |
DOK6 polyclonal antibody detects endogenous levels of DOK6 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only