Learn More
DPP10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DPP10.
Manufacturer:  Abnova Corporation PAB16462
Catalog No. 89-118-334
Add to cart
Description
Description
This gene encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Mutations in this gene have been associated with asthma. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
Specifications
DPP10
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
DPP10
Liquid
DPP10
A synthetic peptide (conjugated with KLH) corresponding to human DPP10.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Extracellular domain of human.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of DPP10.
In PBS (0.09% sodium azide)
Q8N608
DPL2|DPPY|DPRP3
Rabbit
Immunoaffinity purification
RUO
Primary
57628
Bovine, Equine, Human, Monkey, Porcine, Rabbit