Learn More
DPYSL2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DPYSL2.
Supplier:  Abnova Corporation PAB5324
Catalog No. 89-131-817
Add to Cart
Description
Description
Specifications
Specifications
DPYSL2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of DPYSL2.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
CRMP2/DHPRP2/DRP-2/DRP2
Rabbit
100 μg
Primary
Human, Mouse, Rat
Liquid
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
DPYSL2
DPYSL2
A synthetic peptide corresponding to C-terminus of human DPYSL2.
RUO
1808
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations