Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
DRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DRD1.
Manufacturer:  Abnova Corporation PAB12318
Catalog No. 89-115-712
Description
Description
This gene encodes the D1 subtype of the dopamine receptor. The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene. [provided by RefSeq
Specifications
Specifications
DRD1
Unconjugated
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
DRD1
DADR|DRD1A
Rabbit
IgG
100 ug
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL
Polyclonal
Identical to the related rat and mouse sequence.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of DRD1.
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Lyophilized
DRD1
A synthetic peptide corresponding to amino acids at C-terminus of human DRD1.
Affinity purification
RUO
Primary
1812
Human, Murine, Rat