Learn More
ECS, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptides of ECS.
Manufacturer:  Abnova Corporation PAB17058
Catalog No. 89-138-269
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
ECS
Unconjugated
ELISA (2-10 ug/mL) Immunohistochemistry (0.5-5 ug/mL) Immunocytochemistry (0.5-5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide (conjugated with KLH) corresponding to enterokinase cleavage site (XXXDDDK). XXX represents random amino acids.
RUO
Primary
Not Provided
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptides of ECS.
In PBS
Rabbit
1 mg
Store at 4°C.
Polyclonal