Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-EGF, Alexa Fluor 647, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP119806AF647
Description
Specifically detects EGF in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
EGF Polyclonal antibody specifically detects EGF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EGF | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Affinity Purified | |
EGF | |
Synthetic peptide made to an internal portion of the human EGF protein (between amino acids 700-800) [UniProt P01133] | |
Immunogen affinity purified | |
RUO | |
Store at 4C in the dark. | |
Polyclonal | |
Human, Mouse, Rat |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P01133 | |
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG | |
Rabbit | |
IgG | |
0.1 ml | |
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
Primary | |
1950 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only