Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of EGLN1.

Manufacturer:  Abnova Corporation PAB14555

Catalog No. 89-117-139


Add to cart

Description

Description

Specifications

Specifications

EGLN1
Unconjugated
Flow Cytometry (3 mg/mL) Western Blot (1:500-1:2500) The optimal working dilution should be determined by the end user.
EGLN1
Liquid
EGLN1
A synthetic peptide corresponding to amino acids 1-50 of human EGLN1.
RUO
Primary
54583
Human
Flow Cytometry, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of EGLN1.
In Tris-citrate/phosphate buffer, pH 7-8 (0.09% sodium azide)
NP_071334.1
C1orf12|DKFZp761F179|ECYT3|HIFPH2|HPH2|PHD2|SM-20|SM20|ZMYND6
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody is specific to human HIF prolyl hydroxylase 2.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only