Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EHF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238896
Description
EHF Polyclonal specifically detects EHF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EHF | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9NZC4 | |
EHF | |
This antibody was developed against a recombinant protein corresponding to amino acids: MILEGGGVMNLNPGNNLLHQPPAWTDSYSTCNVSSGFFGGQWHEIHPQYWTKYQVWEWLQHLLDTNQLDANCIPFQEFDINGEHL | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
26298 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Epithelium-specific Ets transcription factor 3, ESE-3, ESE3 transcription factor, ESE3B, ESE3hEHF, ESEJepithelium-specific ets factor 3, ETS domain-containing transcription factor, ets homologous factor | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction