Learn More
EIF2C2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EIF2C2.
Supplier: Abnova Corporation PAB18863
Description
This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
EIF2C2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of EIF2C2. | |
Lyophilized from buffer containing 0.02% sodium azide | |
NM_012154 | |
EIF2C2 | |
A synthetic peptide corresponding to 14 amino acids at C-terminus of human EIF2C2. | |
RUO | |
27161 | |
Store at 4°C on dry atmosphere. After reconstitution with 200μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:80000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
EIF2C2 | |
AGO2/MGC3183/Q10 | |
Rabbit | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only