Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Endoglin/CD105, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP1354710.1ML

Catalog No. NBP13547101

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Endoglin/CD105
Unconjugated
7.0 ml prediluted antibody with 10mM PBS (pH 7.4) and 1.0% BSA with 0.1% Sodium Azide
Affinity Purified
ENG
Recombinant protein encoding 26-176 aa of human CD105
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50
P17813
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Rabbit
IgG
0.1 ml
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
2022
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit