Learn More
Rabbit anti-ENT1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241313
Catalog No. NBP241313
Add to cart
Description
Description
ENT1 Polyclonal antibody specifically detects ENT1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
Specifications
ENT1
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 ug/ml
Affinity Purified
SLC29A1
Antibody was raised against a 17 amino acid peptide near the center of human SLC29A1. The immunogen is located within amino acids 240 - 290 of SLC29A1.
53 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
SLC29A1 antibody is human specific. SLC29A1 antibody is predicted to not cross-react with other SLC29 proteins.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.02% Sodium Azide
ENT1es-type, equilibrative nucleoside transporter 1, MGC1465, MGC3778, solute carrier family 29 (nucleoside transporters), member 1, Solute carrier family 29 member 1, solute carrier family 29, member 1
Rabbit
IgG
Peptide affinity purified
RUO
Primary
2030
Human