Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCAM Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of EPCAM.

Manufacturer:  Abnova Corporation PAB24879

Catalog No. 89-126-551


Add to cart

Description

Description

This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy. [provided by RefSeq

Specifications

Specifications

EPCAM
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of EPCAM.
In 1X PBS, pH 7.2 (0.05% sodium azide)
Liquid
EPCAM
A synthetic peptide corresponding to EPCAM.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
polyclonal antibody detects endogenous levels of EPCAM protein.
Immunofluorescence, Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
EPCAM
17-1A|323/A3|CD326|CO-17A|CO17-1A|EGP|EGP-2|EGP34|EGP40|ESA|Ep-CAM|GA733-2|HEA125|KS1/4|KSA|M4S1|MH99|MIC18|MK-1|MOC31|TACST-1|TACSTD1|TROP1|hEGP-2
Rabbit
Antigen affinity purification
RUO
Primary
4072
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only