Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-EPHA5, Polyclonal, Abnova™

Manufacturer:  Abnova Corporation PAB14575

Catalog No. 89-117-156

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

EPHA5
Unconjugated
NP_004430
EPHA5
A synthetic peptide corresponding to amino acids 157-172 of human EPHA5.
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
ELISA
In serum (50% glycerol, 0.05% sodium azide)
Serum
Rabbit
100μL
Primary
2044
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only