Learn More
EPHX2 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of EPHX2.
Manufacturer: Abnova Corporation PAB25596
Description
This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Specifications
EPHX2 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
EPHX2 | |
CEH|SEH | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
2053 | |
Human, Monkey |
ELISA, Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of EPHX2. | |
In PBS (0.09% sodium azide) | |
Liquid | |
EPHX2 | |
A synthetic peptide corresponding to 17 amino acids at C-terminus of human EPHX2. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only