Learn More
EPS15 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EPS15.
Manufacturer:  Abnova Corporation PAB12437
Catalog No. 89-115-811
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a protein that is part of the EGFR pathway. The protein is present at clatherin-coated pits and is involved in receptor-mediated endocytosis of EGF. Notably, this gene is rearranged with the HRX/ALL/MLL gene in acute myelogeneous leukemias. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Specifications
Specifications
EPS15
Unconjugated
In PBS (0.1% BSA, 0.04% sodium azide)
P42566
AF-1P, AF1P, MLLT5
Rabbit
100μL
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
Immunofluorescence, Immunoprecipitation, Western Blot
epidermal growth factor receptor pathway substrate 15
EPS15
Liquid
EPS15
A synthetic peptide corresponding to amino acids 840-896 of human EPS15.
RUO
Primary
2060