Learn More
ERO1L, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERO1L.
Manufacturer:  Abnova Corporation PAB12432
Catalog No. 89-115-806
Add to cart
Description
Description
Specifications
Specifications
ERO1L
Unconjugated
Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
ERO1L
Liquid
ERO1L
A synthetic peptide corresponding to N-terminus of human ERO1L.
RUO
Primary
30001
Immunocytochemistry, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of ERO1L.
In BBS (1% BSA)
Q96HE7
ERO1-alpha
Rabbit
100 uL
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat