Learn More
ESR1 (phospho S102), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ESR1.
Supplier:  Abnova Corporation PAB29243
Catalog No. 89-130-679
Add to Cart
Description
Description
This gene encodes an estrogen receptor, a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters. [provided by RefSeq
Specifications
Specifications
ESR1
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ESR1.
In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
P03372
ESR1
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S102 of human ESR1.
100 μL
Primary
This antibody detects endogenous levels of ESR1 only when phosphorylated at serine 102.
Store at -20°C.
Avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user.
ESR1
DKFZp686N23123/ER/ESR/ESRA/Era/NR3A1
Rabbit
Affinity purification
RUO
2099
Human, Mouse, Rat
Liquid
Recommendations
Recommendations