Learn More
ETFDH, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ETFDH.
Supplier: Abnova Corporation PAB23884
Description
Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67kDa precursor which is targeted to mitochondria and processed in a single step to a 64kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia. [provided by RefSeq
Sequence: LAVAHEKDIRVCLVEKAAQIGAHTLSGACLDPGAFKELFPDWKEKGAPLNTPVTEDRFGILTEKYRIPVPILPGLPMNSpecifications
ETFDH | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant ETFDH. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q16134 | |
ETFDH | |
Recombinant protein corresponding to amino acids of human ETFDH. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
ETFDH | |
ETFQO/MADD | |
Rabbit | |
Antigen affinity purification | |
RUO | |
2110 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only