Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exosc7, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant Exosc7.

Manufacturer:  Abnova Corporation PAB15754

Catalog No. 89-117-818


Add to cart

Description

Description

Specifications

Specifications

Exosc7
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Exosc7
Liquid
Exosc7
Recombinant GST fusion protein corresponding to 126 amino acids of mouse Exosc7.
RUO
Primary
66446
Human, Murine
Western Blot
Rabbit polyclonal antibody raised against partial recombinant Exosc7.
In PBS (50% glycerol, 0.02% sodium azide)
AK129057
2610002K22Rik|AV212732|mKIAA0116
Rabbit
50 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Specific to recombinant protein GX0677. This antibody detects mEXOSC7 protein. It also recognizes human EXOSC7 protein.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only