Learn More
EZR (phospho T566), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EZR.
Manufacturer: Abnova Corporation PAB25825
Description
The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene. [provided by RefSeq
Sequence: Y-K-Tp-L-RSpecifications
EZR | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EZR. | |
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P15311 | |
CVIL|CVL|DKFZp762H157|FLJ26216|MGC1584|VIL2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Primary | |
7430 |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
EZR | |
Liquid | |
EZR | |
Synthetic phosphopeptide corresponding to residues surrounding T566 of human EZR. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine, Rat |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only