Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-FAM115A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243827

Catalog No. NBP243827


Add to cart

Description

Description

Specifically detects FAM115A in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence

FAM115A Polyclonal antibody specifically detects FAM115A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

FAM115A
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
family with sequence similarity 115, member A, hypothetical protein LOC9747, KIAA0738FLJ56782
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
9747
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1:100-1:1000
Ion Exchange Chromatography
FAM115A
Recombinant protein encompassing a sequence within the C-terminus region of human FAM115A. The exact sequence is proprietary.
102 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only