Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL20 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of FBXL20.

Manufacturer:  Abnova Corporation PAB13325

Catalog No. 89-116-575


Add to cart

Description

Description

Members of the F-box protein family, such as FBXL20, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Specifications

Specifications

FBXL20
Unconjugated
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
FBXL20
Liquid
FBXL20
A synthetic peptide corresponding to C-terminus 18 amino acids of human FBXL20.
RUO
Primary
84961
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of FBXL20.
In PBS (0.02% sodium azide)
NP_116264
Fbl2|Fbl20|MGC15482|SCR|SCRAPPER
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only