Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCGR2B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FCGR2B.
Supplier: Abnova Corporation PAB17294
Specifications
FCGR2B | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of FCGR2B. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
P31994 | |
FCGR2B | |
A synthetic peptide corresponding to residues surrounding serine 363 of human FCGR2B. | |
RUO | |
2213 | |
Human | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
FCGR2B | |
CD32/CD32B/FCG2/FCGR2/IGFR2 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total FCGR2B protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction