Learn More
FDPS, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FDPS.
Supplier:  Abnova Corporation PAB4323
Catalog No. 89-131-364
Add to Cart
Description
Description
This gene encodes an enzyme that catalyzes the production of geranyl pyrophosphate and farnesyl pyrophosphate from isopentenyl pyrophosphate and dimethylallyl pyrophosphate. The resulting product, farnesyl pyrophosphate, is a key intermediate in cholesterol and sterol biosynthesis, a substrate for protein farnesylation and geranylgeranylation, and a ligand or agonist for certain hormone receptors and growth receptors. Drugs that inhibit this enzyme prevent the post-translational modifications of small GTPases and have been used to treat diseases related to bone resorption. Multiple pseudogenes have been found on chromosomes 1, 7, 14, 15, 21 and X. Multiple transcript variants encoding different isoforms have been found for this gene
Specifications
Specifications
FDPS
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of FDPS.
In PBS (0.09% sodium azide)
FPPS_HUMAN
FDPS
A synthetic peptide (conjugated with KLH) corresponding to amino acids 338-453 at internal region of human FDPS.
400 μL
Primary
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
FDPS
FPPS/FPS
Rabbit
Protein G purification
RUO
2224
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions