Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF7 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant FGF7.

Manufacturer:  Abnova Corporation PAB17966

Catalog No. 89-119-584


Add to cart

Description

Description

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is a potent epithelial cell-specific growth factor, whose mitogenic activity is predominantly exhibited in keratinocytes but not in fibroblasts and endothelial cells. Studies of mouse and rat homologs of this gene implicated roles in morphogenesis of epithelium, reepithelialization of wounds, hair development and early lung organogenesis. [provided by RefSeq

Specifications

Specifications

FGF7
Unconjugated
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user.
FGF7
Liquid
FGF7
Recombinant protein corresponding to human FGF7.
RUO
Primary
2252
ELISA, Inhibition Assays, Western Blot
Rabbit polyclonal antibody raised against recombinant FGF7.
In PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal)
NP_002000
HBGF-7|KGF
Rabbit
100 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only