Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt1, Rabbit, DyLight 488, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Flt1.
Supplier: Abnova Corporation PAB15091
Specifications
Flt1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Flt1. | |
In 50mM sodium borate | |
AI323757/Flt-1/VEGFR-1/VEGFR1/sFlt1 | |
Rabbit | |
100 μg | |
Primary | |
Other species have not yet been tested. This antibody has a high likelihood of also reacting with mouse and rat proteins. This has only been shown to recognize the native form (180-190 KDa protein) of VEGFR-2, known as Flk-1 / KDR / VEGFR 2 (Vascular Endothelial Growth Factor Receptor 2), though it was raised against the precursor form. This antibody is useful for Western blot analysis, where a doublet is seen at ∼150 KDa with CSF-1R/VEGFR2 chimera transfected lysates representing VEGFR-2. A non-specific band is also seen at ∼85 KDa. | |
Store at 4°C. Do not freeze. |
Flow Cytometry, Western Blot | |
DyLight 488 | |
Flow Cytometry (1:100) The optimal working dilution should be determined by the end user. | |
Flt1 | |
Flt1 | |
A synthetic peptide corresponding to amino acids 800-900 of mouse Flt1. | |
RUO | |
14254 | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction