Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt1, Rabbit, DyLight 649, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Flt1.
Supplier: Abnova Corporation PAB15089
Specifications
Flt1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Flt1. | |
In 50mM sodium borate | |
AI323757/Flt-1/VEGFR-1/VEGFR1/sFlt1 | |
Rabbit | |
100 μg | |
Primary | |
This is specific for VEGFR-1 with significant cross-reactivity with the VEGFR-2 protein. | |
Store at 4°C. Do not freeze. |
Flow Cytometry, Western Blot | |
DyLight 649 | |
Flow Cytometry (1:100) The optimal working dilution should be determined by the end user. | |
Flt1 | |
Flt1 | |
A synthetic peptide corresponding to amino acids 800-900 of mouse Flt1. | |
RUO | |
14254 | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction