Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLT4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant FLT4.

Manufacturer:  Abnova Corporation PAB16189

Catalog No. 89-118-079


Add to cart

Description

Description

This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA. [provided by RefSeq

Specifications

Specifications

FLT4
Unconjugated
ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
FLT4
FLT41|LMPH1A|PCL|VEGFR3
Rabbit
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human
ELISA
Rabbit polyclonal antibody raised against recombinant FLT4.
Lyophilized from PBS.
Lyophilized
FLT4
Recombinant protein corresponding to human FLT4.
RUO
Primary
2324
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only