Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fn14, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant Fn14.

Manufacturer:  Abnova Corporation PAB13804

Catalog No. 89-137-687


Add to cart

Description

Description

Specifications

Specifications

Fn14
Unconjugated
Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Recombinant human IgG1 Fc fusion protein corresponding to Fn14.
RUO
Primary
Recognizes Fn14.
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against recombinant Fn14.
In PBS (0.02% sodium azide)
Rabbit
100 ug
Store at 4°C.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only