Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FTO Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060935AF405
Description
FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.Specifications
FTO | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunoblotting | |
Q9C0B1 | |
FTO | |
A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Chicken, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunoblot | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
Rabbit | |
60 kDa | |
0.1 mL | |
Lipid and Metabolism, Neuroscience | |
79068 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction