Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTO, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of FTO.

Manufacturer:  Abnova Corporation PAB16736

Catalog No. 89-118-572


Add to cart

Description

Description

The exact function of this gene is not know. Studies in mice suggest that it may be involved in nucleic acid demethylation, and that its mRNA level is regulated by feeding and fasting. Genomewide association studies of type 2 diabetes indicate this gene as a diabetes susceptibility locus. Mutation in this gene has been associated with growth retardation, developmental delay, coarse facies, and early death. [provided by RefSeq

Specifications

Specifications

FTO
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
FTO
Liquid
FTO
A synthetic peptide corresponding to N-terminus 15 amino acids of human FTO.
RUO
Primary
79068
ELISA, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of FTO.
In PBS (0.02% sodium azide)
Q9C0B1
KIAA1752|MGC5149
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only