Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUT4, Rabbit, Clone: SP159, Abnova™
Rabbit monoclonal antibody raised against FUT4.
Supplier: Abnova Corporation MAB11048
Description
The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). [provided by RefSeq
Specifications
FUT4 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user. | |
FUT4 | |
FUT4 | |
Lewis-X purified from U937 cells. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (Paraffin) | |
SP159 | |
Rabbit monoclonal antibody raised against FUT4. | |
In PBS, pH 7.6 (1% BSA, 0.09% sodium azide) | |
CD15/ELFT/FCT3A/FUC-TIV/FUTIV | |
Rabbit | |
Protein A/G purification | |
RUO | |
2526 | |
Store at 4°C. Do not freeze. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction