Learn More
FZD9 Rabbit anti-Dog, Human, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.
Supplier: Abnova Corporation PAB26216
Description
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney. [provided by RefSeq
Specifications
FZD9 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of FZD9. | |
In PBS (0.09% sodium azide) | |
O00144 | |
FZD9 | |
A synthetic peptide corresponding to 14 amino acid at N-terminus of human FZD9. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (14 ug/mL) The optimal working dilution should be determined by the end user. | |
FZD9 | |
CD349/FZD3 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
8326 | |
Canine, Human, Rabbit | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only