Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
FZD9 Rabbit anti-Bovine, Dog, Hamster, Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.
Manufacturer:  Abnova Corporation PAB26260
Catalog No. 89-127-900
Description
Description
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney. [provided by RefSeq
Specifications
Specifications
FZD9
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
FZD9
Liquid
FZD9
A synthetic peptide corresponding to 14 amino acid at extracellular domain of human FZD9.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.
In PBS (0.09% sodium azide)
O00144
CD349|FZD3
Rabbit
Immunoaffinity chromatography
RUO
Primary
8326
Bovine, Canine, Hamster, Human, Murine, Rabbit, Rat