Learn More
FZD9 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.
Supplier:  Abnova Corporation PAB26261
Catalog No. 89-127-901
Add to Cart
Description
Description
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney. [provided by RefSeq
Specifications
Specifications
FZD9
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.
In PBS (0.09% sodium azide)
O00144
FZD9
A synthetic peptide corresponding to 14 amino acid at N-terminus of human FZD9.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
FZD9
CD349/FZD3
Rabbit
Immunoaffinity chromatography
RUO
8326
Human, Mouse
Liquid
Product Suggestions
Product Suggestions