Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB1 (phospho Y627), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of GAB1.

Supplier:  Abnova Corporation PAB25870

Catalog No. 89-127-500


Add to Cart

Description

Description

The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family. It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: V-E-Yp-L-D
Specifications

Specifications

GAB1
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
GAB1
GAB1
Synthetic phosphopeptide corresponding to residues surrounding Y627 of human GAB1.
100 μg
Primary
Human, Mouse
Liquid
Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GAB1.
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Q13480
Rabbit
Affinity chromatography
RUO
2549
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only