Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236560

Catalog No. NBP236560


Add to cart

Description

Description

GABA-AR alpha 2 Polyclonal antibody specifically detects GABA-AR alpha 2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GABA-AR alpha 2
Unconjugated
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Specific for the ~51k alpha2-subunit of the GABAA receptor in Western blots. Labeling is absent in alpha2-subunit knockout animals.
Western Blot, Immunohistochemistry
Western Blot 1:1000, Immunohistochemistry
Affinity Purified
GABRA2
Fusion protein from the cytoplasmic loop of the alpha 2 subunit
51 kDa
0.1 ml
Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
2555
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit