Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gabbr2 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Gabbr2.
Supplier: Abnova Corporation PAB14688
Specifications
Gabbr2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Gabbr2. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
GABABR2/Gb2/Gm425/Gpr51 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes C-terminus of gamma-aminobutyric acid (GABA) B receptor 2 (recognized epitope: the last 23 aa). GB2 apparent MW ∼120 KDa. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Western Blot (0.6 ug/mL) The optimal working dilution should be determined by the end user. | |
Gabbr2 | |
Gabbr2 | |
A synthetic peptide (conjugated with THG) corresponding to C-terminus last 23 amino acids of mouse Gabbr2. | |
RUO | |
242425 | |
Mouse, Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction