Learn More
GAP43 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GAP43.
Supplier: Abnova Corporation PAB5367
Description
The protein encoded by this gene has been termed a 'growth' or 'plasticity' protein because it is expressed at high levels in neuronal growth cones during development and axonal regeneration. This protein is considered a crucial component of an effective regenerative response in the nervous system. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
GAP43 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GAP43. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
B-50/PP46 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
GAP43 | |
GAP43 | |
A synthetic peptide corresponding to residues surrounding S41 of human GAP43. | |
RUO | |
2596 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only